Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BANP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561452
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561452 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561452 Supplier Invitrogen™ Supplier No. PA561452
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PQALHYALAN VQQVQIHQIG EDGQVQ Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

This gene encodes a protein that binds to matrix attachment regions. The protein forms a complex with p53 and negatively regulates p53 transcription, and functions as a tumor suppressor and cell cycle regulator. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BANP
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BANP
Gene Accession No. Q8N9N5
Gene Alias AA408158; Banp; BEN domain-containing protein 1; BEND1; BTG3 associated nuclear protein; Btg3-associated nuclear protein; Protein BANP; scaffold/matrix-associated region-1-binding protein; Smar1; SMARBP1
Gene Symbols BANP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BANP. Recombinant protein control fragment (Product #RP-100362).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54971
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.