Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BANP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598780
Description
BANP Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes a protein that binds to matrix attachment regions. The protein forms a complex with p53 and negatively regulates p53 transcription, and functions as a tumor suppressor and cell cycle regulator. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
BANP | |
Polyclonal | |
Unconjugated | |
BANP | |
AA408158; Banp; BEN domain-containing protein 1; BEND1; BTG3 associated nuclear protein; Btg3-associated nuclear protein; Protein BANP; scaffold/matrix-associated region-1-binding protein; Smar1; SMARBP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54971 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1.01 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8N9N5 | |
BANP | |
Recombinant Human Protein BANP protein (1-260AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction