Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAP1 Antibody (1G8) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280589
Description
BAP1 Monoclonal specifically detects BAP1 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
BAP1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 2 μg/mL, Flow Cytometry : 1 ug per million cells | |
BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase), BRCA1-associated protein 1, Cerebral protein 6, cerebral protein-13, cerebral protein-6, DKFZp686N04275, EC 3.4.19.12, FLJ35406, FLJ37180, hucep-6, KIAA0272HUCEP-13, ubiquitin carboxyl-terminal hydrolase BAP1, ubiquitin carboxy-terminal hydrolase, UCHL2 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, Flow Cytometry | |
1G8 | |
Unconjugated | |
Tris-Glycine, 0.15 M NaCl | |
BAP1 | |
Full-length human BAP1 protein expressed and purified from E.coli. [UniProt# Q92560] | |
0.1 mL | |
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Tumor Suppressors | |
8314 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction