Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAP1 Mouse anti-Human, Clone: BAP1/2432, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | BAP1 |
---|---|
Clone | BAP1/2432 |
Concentration | 0.2 mg/ml |
Dilution | ELISA, Protein Array |
Applications | ELISA, Peptide Array |
Description
BAP1 Monoclonal specifically detects BAP1 in Human samples. It is validated for ELISA, Protein Array.Specifications
BAP1 | |
0.2 mg/ml | |
ELISA, Peptide Array | |
Unconjugated | |
Mouse | |
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Tumor Suppressors | |
10 mM PBS with 0.05% BSA | |
8314 | |
IgG2c κ | |
Protein A or G purified |
BAP1/2432 | |
ELISA, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase), BRCA1-associated protein 1, Cerebral protein 6, cerebral protein-13, cerebral protein-6, DKFZp686N04275, EC 3.4.19.12, FLJ35406, FLJ37180, hucep-6, KIAA0272HUCEP-13, ubiquitin carboxyl-terminal hydrolase BAP1, ubiquitin carboxy-terminal hydrolase, UCHL2 | |
Recombinant human BAP1 protein fragment (around aa 191-326) (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title