Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAP1 Mouse anti-Human, Clone: BAP1/2433, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen BAP1
Clone BAP1/2433
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122258
SDP
View Documents
Novus Biologicals
NBP30739220UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

BAP1 Monoclonal specifically detects BAP1 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

BAP1
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Tumor Suppressors
10 mM PBS with 0.05% BSA
8314
IgG
Protein A or G purified
BAP1/2433
Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
Human
BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase), BRCA1-associated protein 1, Cerebral protein 6, cerebral protein-13, cerebral protein-6, DKFZp686N04275, EC 3.4.19.12, FLJ35406, FLJ37180, hucep-6, KIAA0272HUCEP-13, ubiquitin carboxyl-terminal hydrolase BAP1, ubiquitin carboxy-terminal hydrolase, UCHL2
Recombinant human BAP1 protein fragment (around aa 191-326) (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.