Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ BAP1, Mouse, Unlabeled, Clone: 2A2,

Catalog No. MABE1083 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABE1083 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1083 Supplier MilliporeSigma™ Supplier No. MABE1083

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Serine-protein kinase ATM (EC 2.7.11.1; UniProt Q13315; also known as A-T mutated, Ataxia telangiectasia mutated) is encoded by the ATM (also known as AT) gene (Gene ID 472) in human. Ataxia telangiectasia mutated kinase (ATM) and ataxia telangiectasia and Rad3-related kinase (ATR) are related kinases that regulate cell cycle checkpoints and DNA repair. Mutation in the ATM gene results in the autosomal recessive disease ataxia telangiectasia (AT). Known ATM substrates include p53, p95/NBS1, MDM2, Chk2, BRCA1, CtIP, 4E-BP1, and Chk1. The S/TQ sequence constitutes the essential substrates phosphorylation site motif. Hydrophobic amino acids at positions -3 and -1, and negatively charged amino acids at position +1 are positive determinants for substrate phosphorylation, while positively charged residues surrounding the S/TQ are negative determinants. The complex phenotypes of cells derived from patients with AT suggests that ATM has additional cellular substrates. ATM is present as an inactive homodimer or multimer prior to activation. DNA double-stranded breaks induced by ionizing radiation (IR) cause rapid ATM autophosphorylation at Ser1981 in human or the Ser1987 equivalent in mouse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAP1
Applications Western Blot
Classification Monoclonal
Clone 2A2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene BAP1, KIAA0272, hucep-6
Gene Accession No. Q92560
Host Species Mouse
Immunogen Recombinant full-length human BAP1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_004647
Test Specificity Clone 2A2 targets human BAP1 UCH domain.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.