Learn More
Invitrogen™ BAP1 Polyclonal Antibody

Description
Immunogen sequence: EKYSPKELLA LLKCVEAEIA NYEACLKEEV EKRKKFKIDD QRRTHNYDEF ICTFISMLAQ EGMLANLVEQ NISVRRRQGV SIGRLHKQRK P Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
Specifications
Specifications
| Antigen | BAP1 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.10 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | BAP1 |
| Gene Accession No. | Q92560 |
| Gene Alias | 2300006C11Rik; AA989761; AW553466; BAP1; Brca1 associated protein 1; BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase); BRCA1-associated protein 1; Cerebral protein 6; cerebral protein-13; DKFZp686N04275; FLJ35406; FLJ37180; HUCEP-13; hucep-6; KIAA0272; mKIAA0272; Ubiquitin carboxyl-terminal hydrolase BAP1; ubiquitin C-terminal hydrolase X4; UCHL2; uch-x4 |
| Gene Symbols | BAP1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.