Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAP29 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176421MFV450
Description
BAP29 Polyclonal antibody specifically detects BAP29 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
BAP29 | |
Polyclonal | |
Western Blot, ELISA | |
Bap29, BAP29DKFZp686M2086, B-cell receptor-associated protein 29, BCR-associated protein 29, FLJ53907 | |
Antibody was raised against a 17 amino acid synthetic peptide from near the center of human Bap29. Bap29 antibody is specific for isoform a of Bap29. The immunogen is located within amino acids 230 - 280 of BAP29. Amino Acid Squence: KRSHKNGSIGKQTGSRK | |
0.1 mL | |
Apoptosis | |
55973 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction