Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BAP29 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520196
Description
This antibody is specific for isoform A of Bap29. A suggested positive control is human heart tissue lysate. PA5-20196 can be used with blocking peptide PEP-0326.
Bap29 and the related protein Bap31 are endoplasmic reticulum (ER) and ER-vesicle membrane proteins and members of the B-cell receptor-associated protein family. These two proteins are highly homologous and can form homo- and heterodimers. Both Bap29 and Bap31 interact with membrane-bound immunoglobulins (mIgs), such as IgM and IgD, which with Ig-alpha/Ig-beta heterodimers form B cell antigen receptors. Binding of the Bap29/Bap31 heterodimer correlates with the ER retention of non-Ig-alpha/Ig-beta bound mIg complexes, suggesting that Bap29 and Bap31 may act as chaperones transmembrane regions of various proteins. Bap29 possesses multiple isoforms.
Specifications
| BAP29 | |
| Polyclonal | |
| Unconjugated | |
| Bcap29 | |
| AW208404; B cell receptor associated protein 29; BAP29; Bcap29; B-cell receptor-associated protein 29; B-cell receptor-associated protein BAP29; BCR-associated protein 29 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12033, 298943, 55973 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q61334, Q9UHQ4 | |
| Bcap29 | |
| A 17 amino acid synthetic peptide from near the center of human Bap29. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction