Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAP29 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562029
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562029 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562029 Supplier Invitrogen™ Supplier No. PA562029
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLVEDQEKLK TELRKTSDAL SKAQNDVMEM KMQSERLSKE YDQLLKEHSE LQDRLERGNK K Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 82%.

Bap29 and the related protein Bap31 are endoplasmic reticulum (ER) and ER-vesicle membrane proteins and members of the B-cell receptor-associated protein family. These two proteins are highly homologous and can form homo- and heterodimers. Both Bap29 and Bap31 interact with membrane-bound immunoglobulins (mIgs), such as IgM and IgD, which with Ig-alpha/Ig-beta heterodimers form B cell antigen receptors. Binding of the Bap29/Bap31 heterodimer correlates with the ER retention of non-Ig-alpha/Ig-beta bound mIg complexes, suggesting that Bap29 and Bap31 may act as chaperones transmembrane regions of various proteins. Bap29 possesses multiple isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAP29
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Bcap29
Gene Accession No. Q9UHQ4
Gene Alias AW208404; B cell receptor associated protein 29; BAP29; Bcap29; B-cell receptor-associated protein 29; B-cell receptor-associated protein BAP29; BCR-associated protein 29
Gene Symbols Bcap29
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BAP29. Recombinant protein control fragment (Product #RP-103052).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55973
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.