Learn More
Invitrogen™ BAP3 Polyclonal Antibody

Description
A suggested positive control is SK-N-SH cell lysate. PA5-20498 can be used with blocking peptide PEP-0618.
Specifications
Specifications
| Antigen | BAP3 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | BAIAP3 |
| Gene Accession No. | O94812 |
| Gene Alias | AL031709.1; BAI1 associated protein 3; BAI1-associated protein 3; BAIAP3; BAI-associated protein 3; BAP3; Brain-specific angiogenesis inhibitor I-associated protein 3; Gm937; KIAA0734 |
| Gene Symbols | BAIAP3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.