Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAP31 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596563
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596563 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596563 Supplier Invitrogen™ Supplier No. PA596563
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATLLASNEAF KKQAESASEA AKKYMEENDQ LKKGAAVDGG KLDVGNAEVK LEEENRSLKA DLQKLKDELA STKQKLEKAE NQVLAMRKQS EGLTKEYDRL LEEHAKLQAA VDGPMDKKEE; Positive Samples: MCF-7, HT-29, SH-SY5Y, HeLa, A-431, Mouse liver, Mouse kidney, Mouse lung, Rat brain; Cellular Location: Endoplasmic reticulum membrane, Endoplasmic reticulum-Golgi intermediate compartment membrane, Multi-pass membrane protein.

BAP31 (B-Cell Receptor Associated Protein, 31-kDa) is a multiple membrane spanning protein of the endoplasmic reticulum. It has been shown to form both homo- and hetero-oligomers with the closely related BAP29, and is a potential apoptotic regulator as a predicted BCL-2/BCL-XL-associated protein. In the C-terminal domain of BAP31, there are two identical recognition sites for initiator Caspases 1 and 8 of the programmed death cascade. It is hypothesized that BAP31 plays a role in the transport of selected proteins to the Golgi apparatus. BAP31 has also been shown to control expression of CFTR (cystic fibrosis transmembrane conductance regulator). It is believed that interference with the expression or function of BAP31 in epithelial cells may be a way to circumvent the chloride channel defect in cystic fibrosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAP31
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BCAP31
Gene Accession No. P51572, Q61335
Gene Alias 6C6 antigen; 6C6-AG; 6C6-AG tumor-associated antigen; accessory protein BAP31; B cell receptor associated protein 31; BAP31; Bcap31; B-cell receptor-associated protein 31; BCR-associated protein 31; BCR-associated protein Bap31; CDM; CDM protein; DDCH; DXS1357E; hypothetical protein LOC533949; OTTHUMP00000025978; p28; p28 Bap31; Protein CDM
Gene Symbols BCAP31
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 127-246 of human BAP31 (NP_0012433761).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10134, 27061, 293852
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.