Learn More
BAPTA, 97%
Description
Acts as a calcium chelatorBAPTA is used as a selective calcium chelating reagent. It is suitable for the spectrophotometric monitoring of calcium levels. It is a lipophilic diesters of the chelating agent used for the inhibition of proteolytic activities of certain metalloproteinases, calpain and transcatheter arterial chemoembolization (TACE).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acts as a calcium chelatorBAPTA is used as a selective calcium chelating reagent. It is suitable for the spectrophotometric monitoring of calcium levels. It is a lipophilic diesters of the chelating agent used for the inhibition of proteolytic activities of certain metalloproteinases, calpain and transcatheter arterial chemoembolization (TACE).
Solubility
Soluble in water and sodium bicarbonate.
Notes
Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | ∼174°C (decomposition) |
Quantity | 5 g |
Merck Index | 14,957 |
Solubility Information | Soluble in water and sodium bicarbonate. |
Formula Weight | 476.44 |
Percent Purity | 97% |
Chemical Name or Material | BAPTA |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
RTECSNumber | MC0423000 |
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.