Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BAPTA AM
GREENER_CHOICESDP

Catalog No. 278725 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-872-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 27-872-5 Supplier Tocris Bioscience™ Supplier No. 2787/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cell-permeable Ca2+ chelator

Selective calcium chelator that is the cell-permeable analog of BAPTA (Cat. No. 2786). Blocks hKv1.5, hKV11.1 (hERG) and hKv1.3 channels (Ki values are 1.23, 1.30 and 1.45 μM respectively). Displays antithrombotic activity in vitro.

F 127 (Cat. No. 6253) for the solubilization of BAPTA AM is also available.

Chemical Identifiers

CAS 126150-97-8
Molecular Formula C34H40N2O18

Specifications

Quantity 25 mg
Formula Weight 764.68
Purity >95%
Molecular Weight (g/mol) 764.69
Chemical Name or Material BAPTA AM
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.