Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAPTA tetramethyl ester, 99%

Catalog No. AAH6690706
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 125367-34-2 | C26H32N2O10 | 532.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125367-34-2
Molecular Formula C26H32N2O10
Molecular Weight (g/mol) 532.55
MDL Number MFCD00532664
InChI Key LCPLRKMZANZSAJ-UHFFFAOYSA-N
Synonym bapta-tetramethyl ester, tetramethyl 2,2',2,2'-ethane-1,2-diylbis oxy bis 2,1-phenylene bis azanetriyl tetraacetate, tetramethyl 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetate, methyl 2-2-2-2-bis 2-methoxy-2-oxoethyl amino phenoxy ethoxy phenyl 2-methoxy-2-oxoethyl amino acetate, 1,2-bis 2-bis 2-oxo-2-methoxyethyl amino phenoxy ethane, tetramethyl 1,2-bis 2-aminophenoxy ethane-n,n,n,n-tetraacetate, 1,2-bis 2-aminophenoxy ethane-n,n,n,n-tetraacetic acid, tetramethyl ester, glycine, n,n'-1,2-ethanediylbis oxy-2,1-phenylene bis n-2-methoxy-2-oxoethyl-, 1,1'-dimethyl ester, methyl 2-2-2-2-bis 2-methoxy-2-oxoethyl amino phenoxy ethoxy-n-2-methoxy-2-oxoethyl anilino acetate, n,n'-1,2-ethanediylbis oxy-2,1-phenylene bis n-2-methoxy-2-oxoethyl glycine 1,1'-dimethyl ester
PubChem CID 3452402
IUPAC Name methyl 2-{[2-(2-{2-[bis(2-methoxy-2-oxoethyl)amino]phenoxy}ethoxy)phenyl](2-methoxy-2-oxoethyl)amino}acetate
SMILES COC(=O)CN(CC(=O)OC)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC(=O)OC)CC(=O)OC

Specifications

Color White
Quantity 5 g
Formula Weight 532.55
Percent Purity 99%
Physical Form Powder
Chemical Name or Material BAPTA tetramethyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.