Learn More
Aspen Surgical™ Bard-Parker™ Surgical Blades
Catalog No. 0891513
Click to view available options
Size:
# 10
# 11
# 15
# 20
# 21
# 22
# 23
# 25
Sterility:
Non-sterile
Sterile
Handle Type:
Ribbed
Safetylock Ribbed
Offer consistent sharpness, control and strength
  • Each blade packaged in foil pouch with liner for maximum protection of blade and user
  • Rib back blade design on select blades offers greater cutting control and strength
  • Available with plastic shield SafetyLock to securely cover the blade edge before and after use
  • Available sterile in carbon and stainless steel and nonsterile in carbon steel
Specifications
Blade Material Carbon Steel
Product Type Surgical Blade
For Use With (Application) For Tissue Separation, and Other Procedures that require a Sharp Surgical Blade to Puncture or Cut
Handle Type Safetylock Ribbed
Size # 11
Sterility Sterile
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.