Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BARD1 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100319AF647
Description
BARD1 Polyclonal antibody specifically detects BARD1 in Human samples. It is validated for Western BlotSpecifications
BARD1 | |
Polyclonal | |
Western Blot | |
BARD-1, BRCA1 associated RING domain 1, BRCA1-associated RING domain gene 1, BRCA1-associated RING domain protein 1, EC 6.3.2.- | |
The immunogen recognized by this antibody maps to a region between residues 250 and 300 of human BRCA1-associated RING Domain 1 using the numbering given in entry NP_000456.1 (GeneID 580). | |
0.1 mL | |
Breast Cancer, Cancer | |
580 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction