Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BARD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560850
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560850 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560850 Supplier Invitrogen™ Supplier No. PA560850
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MIQLCSKLRN LLHDNELSDL KEDKPRKSLF NDAGNKKNSI KMWFSPRSKK VRYVVSKASV QTQPAIKKDA SAQQDS Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 67%.

This gene encodes a protein which interacts with the N-terminal region of BRCA1. In addition to its ability to bind BRCA1 in vivo and in vitro, it shares homology with the 2 most conserved regions of BRCA1: the N-terminal RING motif and the C-terminal BRCT domain. The RING motif is a cysteine-rich sequence found in a variety of proteins that regulate cell growth, including the products of tumor suppressor genes and dominant protooncogenes. This protein also contains 3 tandem ankyrin repeats. The BARD1/BRCA1 interaction is disrupted by tumorigenic amino acid substitutions in BRCA1, implying that the formation of a stable complex between these proteins may be an essential aspect of BRCA1 tumor suppression. This protein may be the target of oncogenic mutations in breast or ovarian cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BARD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BARD1
Gene Accession No. Q99728
Gene Alias BARD 1; Bard1; BARD-1; BRCA1 associated RING domain 1; BRCA1-associated RING domain gene 1; BRCA1-associated RING domain protein 1; ENSMUSG00000060893; ENSMUSG00000073653; RING-type E3 ubiquitin transferase BARD1
Gene Symbols BARD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BARD1. Recombinant protein control fragment (Product #RP-96098).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 580
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.