Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BARHL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$697.00
Specifications
Antigen | BARHL1 |
---|---|
Concentration | 0.7mg/mL |
Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
BARHL1 Polyclonal antibody specifically detects BARHL1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
BARHL1 | |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
56751 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LELSPRSESSSDCSSPASPGRDCLETGTPRPGGASGPGLDSHLQPGQLSAPAQSRTVTSSFLIRDILADCKPLAACAPYSSSGQPAAPEPGGRLAAKAAEDFRDKLDKSGSNASSDSEYKVKEEGDREISSSRDSP | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.7mg/mL | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat | |
BarH (Drosophila)-like 1, barH-like 1 homeobox protein, BarH-like homeobox 1 | |
BARHL1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title