Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BARHL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552114
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552114 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552114 Supplier Invitrogen™ Supplier No. PA552114
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LELSPRSESS SDCSSPASPG RDCLETGTPR PGGASGPGLD SHLQPGQLSA PAQSRTVTSS FLIRDILADC KPLAACAPYS SSGQPAAPEP GGRLAAKAAE DFRDKLDKSG SNASSDSEYK VKEEGDREIS SSRDSP Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

BARHL1 (BarH Like Homeobox 1) is a Protein Coding gene. Diseases associated with BARHL1 include Deafness, Autosomal Recessive 27 and Deafness, Autosomal Dominant 16. Gene Ontology (GO) annotations related to this gene include sequence-specific DNA binding and proximal promoter DNA-binding transcription activator activity, RNA polymerase II-specific. An important paralog of this gene is BARHL2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BARHL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BARHL1
Gene Accession No. P63157, Q9BZE3
Gene Alias Bar-class homeodomain protein MBH2; BarH like homeobox 1; BARHL1; Barhl2; BarH-like 1 (Drosophila); barH-like 1 homeobox protein; BarH-like 2; BarH-like homeobox 1; barH-related homeobox protein 1; Bhx1; Dres115; FKSG31; Mbh2; PKCdeltaVIII
Gene Symbols BARHL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BARHL1. Recombinant protein control fragment (Product #RP-102508).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54422, 56751
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.