Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BARHL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103365
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103365 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103365 Supplier Invitrogen™ Supplier No. PA5103365
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BARHL2.

BARHL2 and BARHL1 are two homeobox genes in mouse and human, which are highly related to the Bar Drosophila genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BARHL2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BARHL2
Gene Accession No. O88181, Q8VIB5, Q9NY43
Gene Alias bar-class homeodomain protein MBH1; Barh; BarH like homeobox 2; Barh1; BarH-class homeodomain transcription factor; BARHL2; BarH-like 2; BarH-like 2 (Drosophila); barH-like 2 homeobox protein; BarH-like homeobox 2; B-H2; E130309B19Rik; Homeobox protein B-H1; MBH1; wu:fq39g04; zgc:113948
Gene Symbols BARHL2
Host Species Rabbit
Immunogen A synthesized peptide derived from human BARHL2(Accession Q9NY43), corresponding to amino acid residues Q111-S161.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104382, 343472, 65050
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.