Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BARHL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113849
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113849 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113849 Supplier Invitrogen™ Supplier No. PA5113849
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: REITSSRESP PVRAKKPRKA RTAFSDHQLN QLERSFERQK YLSVQDRMDL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

BARHL2 and BARHL1 are two homeobox genes in mouse and human, which are highly related to the Bar Drosophila genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BARHL2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene BARHL2
Gene Accession No. Q9NY43
Gene Alias bar-class homeodomain protein MBH1; Barh; BarH like homeobox 2; Barh1; BarH-class homeodomain transcription factor; BARHL2; BarH-like 2; BarH-like 2 (Drosophila); barH-like 2 homeobox protein; BarH-like homeobox 2; B-H2; E130309B19Rik; Homeobox protein B-H1; MBH1; wu:fq39g04; zgc:113948
Gene Symbols BARHL2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human BARHL2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 343472
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.