Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BARHL2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen BARHL2
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB126744
SDP
View Documents
Novus Biologicals
NBP31092225UL
25 μg
Each for $206.00
Only null left
Add to Cart
 
NB126743
SDP
View Documents
Novus Biologicals
NBP310922100UL
100 μg
Each for $487.50
Only null left
Add to Cart
 
Description

Description

BARHL2 Polyclonal specifically detects BARHL2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

BARHL2
Western Blot, Immunohistochemistry
Unconjugated
Rabbit
Human
BarH (Drosophila)-like 2, barH-like 2 homeobox protein, BarH-like homeobox 2
The immunogen is a synthetic peptide directed towards the middle region of human BARHL2 (NP_064447). Peptide sequence SSPHHTPKQESNAVHESFRPKLEQEDSKTKLDKREDSQSDIKCHGTKEEG
Affinity purified
Western Blot 1.0 ug/ml, Immunohistochemistry
Polyclonal
Purified
RUO
PBS buffer, 2% sucrose
343472
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.