Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Baricitinib
GREENER_CHOICESDP

Catalog No. 722250 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-225-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 72-225-0 Supplier R&D Systems™ Supplier No. 7222/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Highly potent JAK1 and JAK2 inhibitor; also inhibits JAK3 and Tyk2

Baricitinib is a highly potent JAK1 and JAK2 inhibitor (IC50 values are 5.9 and 5.7 nM, respectively). Also inhibits Tyk2 (IC50 = 53 nM). Displays >100-fold selectivity for JAK1/2 over JAK3 (IC50 = 53 nM for JAK3). Binds AAK1, BIKE, GAK and MPSK1 in vitro (Kd values are 17.2, 39.8, 134.4 and 68.5 nM, respectively). Inhibits IL-6-induced STAT phosphorylation and subsequent pro-inflammatory chemokine (MPC-1) and cytokine (IL-17 and IL-22) production in PBMCs and T-cells. Displays anti-inflammatory and disease modifying effects in the rat adjuvant arthritis model. In silico modelling predicts inhibition of SARS-CoV-2 cell entry.

Specifications

Chemical Name or Material 1-(Ethylsulfonyl)-3-[4-(7 H-pyrrolo[2,3-d]pyrimidin-4-yl)-1 H-pyrazol-1-yl]-3-azetidineacetonitrile
CAS 1187594-09-7
Quantity 50 mg
Product Line Tocris
Target JAK Kinase Inhibitors
Molecular Formula C16H17N7O2S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.