Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium bromate, 97%

Catalog No. p-7081159
Click to view available options
Quantity:
100 g
500 g

CAS: 13967-90-3 | BaBr2O6 | 393.129 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13967-90-3
Molecular Formula BaBr2O6
Molecular Weight (g/mol) 393.129
MDL Number MFCD00014177
InChI Key VEASZGAADGZARC-UHFFFAOYSA-L
Synonym barium bromate, bromic acid, barium salt, unii-5qk4064y0r, barium 2+ dibromate, acmc-1bumj, ba.2bro3, bromic acid, barium salt 2:1, barium bromate un2719 oxidizer
PubChem CID 61706
IUPAC Name barium(2+);dibromate
SMILES [O-]Br(=O)=O.[O-]Br(=O)=O.[Ba+2]

Specifications

Quantity 100 g
UN Number UN2719
Formula Weight 393.16
Percent Purity 97%
Odor Odorless
Density 3.99
Chemical Name or Material Barium bromate
Hazard Category H272-H302+H332
Hazard Statement GHS H Statement
H272-H302-H332
May intensify fire
oxidizer.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: BARIUM BROMATE
EINECSNumber 237-750-5
RTECSNumber EF8715000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.