Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium carbonate, 99+%, for analysis

Catalog No. p-45773
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 513-77-9 | CBaO3 | 197.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-77-9
Molecular Formula CBaO3
Molecular Weight (g/mol) 197.34
MDL Number MFCD00003448
InChI Key AYJRCSIUFZENHW-UHFFFAOYSA-L
Synonym barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
PubChem CID 10563
SMILES [Ba++].[O-]C([O-])=O

Specifications

Boiling Point 1450.0°C
Melting Point 811.0°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99% min. (Complexometry)
Linear Formula BaCO3
Merck Index 15, 965
Solubility Information Solubility in water: 0.002g/100mL (20°C)
IUPAC Name barium(2+) carbonate
Formula Weight 197.35
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 4.43
Density 4.4300g/mL
Chemical Name or Material Barium carbonate, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-167-3
RTECSNumber CQ8600000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.