Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium carbonate, Puratronic™, 99.997% (metals basis)

Catalog No. AA1064522
Click to view available options
Quantity:
25 g
100 g
500 g

Catalog No. AA1064522

Supplier: Thermo Scientific Chemicals 01064522

Only null left
Add to Cart

CAS: 513-77-9 | CBaO3 | 197.34 g/mol

Barium carbonate is frequently employed in ceramic industry as a component of glazes. It is added to clay to precipitate some soluble salts while manufacturing bricks, tiles and potteries. Barium carbonate is mainly used as a raw material in the electrical engineering industries for glass production, and as a raw material in the magnet manufacturing industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-77-9
Molecular Formula CBaO3
Molecular Weight (g/mol) 197.34
MDL Number MFCD00003448
InChI Key AYJRCSIUFZENHW-UHFFFAOYSA-L
Synonym barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
PubChem CID 10563
SMILES [Ba++].[O-]C([O-])=O

Specifications

Boiling Point (decomposition)
Melting Point 811°C
Physical Form Lumps, Flakes and Granular Powder
Quantity 100 g
Merck Index 14,969
Solubility Information Insoluble in water. Soluble in most of the acids,with the exception of sulfuric acid.
IUPAC Name barium(2+) carbonate
Formula Weight 197.35
Percent Purity 99.997%
Grade Puratronic
Odor Odorless
Density 4.43 g/mL
Assay (metals basis)
Chemical Name or Material Barium carbonate
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 208-167-3
RTECSNumber CQ8600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.