Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Barium Chloride, 0.100 N (N/10), 0.0500 M (M/20), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
Specifications
Specifications
Color | Colorless |
CAS Min % | 97.0, 1.18 |
CAS Max % | 100.0, 1.23 |
Grade | Laboratory |
Packaging | Natural Poly Bottle |
pH | ∼5 to 7 |
Quantity | 500 mL |
Physical Form | Liquid |
Concentration or Composition Notes | 0.100Normal,0.0500Molar |
Chemical Name or Material | Barium Chloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction