Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Barium Chloride, 0.5 Molar, Unstandardized, Reagents
Click to view available options
Quantity:
4 L
10 L
20 L
Specifications
Specifications
Chemical Name or Material | Barium Chloride, 0.5 Molar, Unstandardized |
Boiling Point | 101°C |
Color | Colorless |
CAS | 7732-18-5, 10326-27-9 |
Physical Form | Liquid |
Quantity | 10 L |
Molecular Formula | BaCl2/H2O |
Grade | Reagent |
Packaging | Cubitainer |
Specific Gravity | 1.086 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction