Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Barium chloride dihydrate, 99+%, for analysis, ACROS Organics™
Click to view available options
Quantity:
500g
1kg
2.5kg
Packaging:
Plastic bottle
Description
For analysis
Specifications
Assay Percent Range | 99% min. (Argentometry) |
Linear Formula | BaCl2·2H2O |
Merck Index | 15, 967 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in methanol,practically insoluble in ethanol,acetone and,ethyl acetate |
Formula Weight | 244.28 |
Physical Form | Metal |
Percent Purity | 99+% |
Grade | Analysis |
Calcium (Ca) | 500ppm max. |
Heavy Metals (as Pb) | 5ppm max. |
Show More |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear eye protection/face protection.
If eye irritation per
GHS Signal Word: Danger