Alfa Aesar™ Barium chloride dihydrate, ACS

Manufacturer: Alfa Aesar™ 12310A3

Quantity 2kg

 View more versions of this product

Catalog No. AA12310A3

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10326-27-9
    Molecular Formula BaCl2·2H2O
    Formula Weight 244.28 (208.25 Anhydrous)
    MDL Number MFCD00149154
    UN Number UN1564
    Merck Index 14,971
    Chemical Name or Material Barium chloride dihydrate, dihydrate
    Grade ACS Grade

    Specifications

    Formula Weight 244.28 (208.25 Anhydrous)
    Melting Point 113°C (in -2H2O)
    Molecular Formula BaCl2·2H2O
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P304+P340-P312-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    UN Number UN1564
    Merck Index 14,971
    Health Hazard 1 Danger
    RTECS Number CQ8751000
    TSCA TSCA
    Physical Form Crystalline
    MDL Number MFCD00149154
    CAS 10326-27-9
    Solubility Information Very soluble in water; Soluble in methanol; Insoluble in ethanol, acetone, ethyl acetate
    Density 3.097g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
    Quantity 2kg
    Health Hazard 2 GHS H Statement
    H301-H332
    Toxic if swallowed.
    Harmful if inhaled.
    Chemical Name or Material Barium chloride dihydrate, dihydrate
    Grade ACS Grade

    GHS H Statement
    H301-H332
    Toxic if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P301+P310-P304+P340-P312-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit