Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Chloride Dihydrate, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCB008025A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Natural Poly Bottle
White Poly Wide Mouth Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCB008025A 2.5 kg Natural Poly Bottle
RDCB0080100 100 g White Poly Wide Mouth Bottle
RDCB0080500 500 g White Poly Wide Mouth Bottle
RDCB00801KG 1 kg White Poly Wide Mouth Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RDCB008025A Supplier Ricca Chemical Company Supplier No. RDCB00802.5A5
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
PubChem CID 5284346
ChEBI CHEBI:86153
IUPAC Name barium(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

Color White
CAS Min % 98.0
CAS Max % 100.0
Physical Form Crystals or Powder
Quantity 2.5 kg
Synonym barium chloride dihydrate
Percent Purity ≥99%
Grade ACS
Loss on Drying 14.0 to 16.0%
Packaging Natural Poly Bottle
Chemical Name or Material Barium Chloride, Dihydrate
Show More Show Less
ShelfLife 60 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.