Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Chloride, Dihydrate, Crystal, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18608986 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-986 500 g Poly Bottle
18-608-985 125 g Poly Bottle
18-608-987 2.5 kg Poly Bottle
18-608-988 12 kg Poly Bottle
18-608-989 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-608-986 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B1025500GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
IUPAC Name barium(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

pH 5.2 to 8.2
CAS Max % 1
Physical Form Crystals and/or Chunks
Quantity 500 g
Percent Purity 99%
Grade Reagent
Loss on Drying 14.0-16.0%
Packaging Poly Bottle
Chemical Name or Material Barium Chloride, Dihydrate, Crystal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.