Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium chloride dihydrate, Puriss. p.a. plus, ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Solstice

Catalog No. 6000360 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
4 x 5 kg
6 x 1 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-003-60 100 g
60-003-63 500 g
60-003-61 1 kg
60-003-64 5 kg
60-045-910 6 x 1 kg
60-045-909 4 x 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-003-60 Supplier Solstice Supplier No. 31125100G
Add to Cart
Add to Cart

Puriss. p.a. plus, ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99%

  • Exceeds standard ACS specifications with additional trace metals analysis
  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PubChem CID 5284346
ChEBI CHEBI:86153
IUPAC Name barium(2+);dichloride;dihydrate
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

Quantity 100 g
UN Number 1564
Formula Weight 244.26g/mol
Percent Purity ≥99%
Grade ACS/puriss. p.a. plus
Packaging Plastic Bottle
Chemical Name or Material Barium Chloride Dihydrate, Reag. Ph. Eur. reag. ISO
Hazard Statement P261
Precautionary Statement H301-H332
EINECSNumber 233-788-1
RTECSNumber CQ8751000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.