Learn More
Barium chloride dihydrate, Puriss. p.a. plus, ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Solstice
Shop All Honeywell Research Chemicals ProductsDescription
- Exceeds standard ACS specifications with additional trace metals analysis
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Specifications
| Quantity | 500 g |
| UN Number | 1564 |
| Formula Weight | 244.26g/mol |
| Percent Purity | ≥99% |
| Grade | ACS/puriss. p.a. plus |
| Packaging | Plastic Bottle |
| Chemical Name or Material | Barium Chloride Dihydrate, Reag. Ph. Eur. reag. ISO |
Safety and Handling
| Hazard Statement | P261 |
| Precautionary Statement | H301-H332 |
| EINECSNumber | 233-788-1 |
| RTECSNumber | CQ8751000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.