Learn More
Barium chloride dihydrate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals
Description
It is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 113°C-2H2O |
Color | White |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.99% (Metal basis) |
Solubility Information | Soluble in water and metanol. Insoluble in ethanol and ethyl acetate. |
Formula Weight | 244.28 (208.25 Anhydrous) |
Odor | Odorless |
Appearance | White powder |
Density | 3.097 g/cm3 |
Show More |
Safety and Handling
Hazard Category | H301-H319-H332 |
Precautionary Statement | P261-P264b-P270-P271-P280i-P301+P310-P304+P340-P305+P351+P338-P312-P330-P501c |
DOTInformation | Hazard Class: 6.1; Packaging Group: III |
EINECSNumber | 600-412-6 |
RTECSNumber | CQ8751000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.