Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Barium Chloride Solution (10% w/w, Neutralized), Fisher Chemical
Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle
Specifications
Specifications
Boiling Point | 100°C |
Melting Point | 0°C |
Color | Red |
pH | 4.0 |
Physical Form | Liquid |
Quantity | 1 L |
Assay Percent Range | 10% w/w |
UN Number | 1564 |
Percent Purity | 9.5 to 10.5% |
Concentration or Composition (by Analyte or Components) | 10% w/w |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction