Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium diphenylamine-4-sulfonate, For Redox Titration, ≥95%, MilliporeSigma™ Supelco™

Catalog No. 111014920 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
5 g
25 g
50 g
Packaging:
Poly Bottle

Chemical Identifiers

CAS 6211-24-1
Molecular Formula C24H20BaN2O6S2
Molecular Weight (g/mol) 633.88
MDL Number MFCD00007497
InChI Key IVCNVXFNTKXMCA-UHFFFAOYSA-L
Synonym 4-(Phenylamino)benzenesulfonic acid barium salt; Diphenylamine-4-sulfonic acid barium salt
IUPAC Name barium(2+) bis(4-(phenylamino)benzene-1-sulfonate)
SMILES [Ba++].[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1.[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Quantity 50 g
Linear Formula (C12H10NO3S)2 Ba
Formula Weight 633.88
Percent Purity ≥95%
Packaging Poly Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.