Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Diphenylamine-4-sulfonate 98.0+%, TCI America™
SDP

Catalog No. D087625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D087625G 25 g
D08761G 1 g
D08765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D087625G Supplier TCI America Supplier No. D087625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6211-24-1
Molecular Formula C24H20BaN2O6S2
Molecular Weight (g/mol) 633.88
MDL Number MFCD00007497
InChI Key IVCNVXFNTKXMCA-UHFFFAOYSA-L
Synonym barium diphenylamine-4-sulfonate, barium diphenylaminesulfonate, benzenesulfonic acid, 4-phenylamino-, barium salt 2:1, barium bis 4-anilinobenzenesulphonate, barium diphenylamine sulfonate, n-phenylsufanilic acid barium, unii-ssj5f79159, diphenylamine-4-sulfonic acid barium salt, n-phenylsufanilic acid, barium salt, 4-anilinobenzenesulfonate; barium 2+
PubChem CID 80342
IUPAC Name barium(2+) bis(4-(phenylamino)benzene-1-sulfonate)
SMILES [Ba++].[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1.[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Color Gray-White
Quantity 25 g
UN Number 1564
Formula Weight 633.88
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Barium Diphenylamine-4-sulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.