Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Barium Diphenylamine Sulfonate, 0.1% w/v for Chlorine, Reagents
Click to view available options
Quantity:
120 mL
500 mL
1 L
Specifications
Specifications
Boiling Point | 100°C |
Color | Colorless to Light Gray/Blue |
Physical Form | Liquid |
Quantity | 500 mL |
Percent Purity | 0.1% |
Grade | Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1.002 |
Chemical Name or Material | Barium Diphenylamine Sulfonate, 0.1% w/v for Chlorine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction