Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium ethoxide, 99.5% (metals basis), 10% w/v in ethanol

Catalog No. p-7050358
Click to view available options
Quantity:
25 mL
100 mL

CAS: 2914-19-4 | C4H10BaO2 | 227.449 g/mol

It is used in nano-materials & thin films; catalysis & synthesis and optics & glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2914-19-4
Molecular Formula C4H10BaO2
Molecular Weight (g/mol) 227.449
MDL Number MFCD00050488
InChI Key GYIWFHXWLCXGQO-UHFFFAOYSA-N
Synonym barium diethanolate, diethoxybarium, barium 2+ ; ethanolate, ethanol, barium salt 9ci, barium 2+ bis ethoxide, barium 2+ ion bis ethoxide, barium ethoxide solution w/v in ethanol trace metals basis 25ml
PubChem CID 6451474
IUPAC Name barium(2+);ethanolate
SMILES CC[O-].CC[O-].[Ba+2]

Specifications

Color Yellow
Linear Formula Ba(OC2H5)2
Quantity 25 mL
UN Number UN3274
Sensitivity Moisture sensitive
Solubility Information Not miscible with water.
Formula Weight 227.42
Percent Purity 99.5%
Concentration or Composition (by Analyte or Components) 10% w/v in ethanol
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Barium ethoxide
Show More Show Less
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H225-H311-H314-H371-H332
Highly flammable liquid and vapor.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause damage to organs.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S.
EINECSNumber 220-829-3
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

For Manufacturing and Laboratory Use. Not for beverage use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.