Learn More
Barium fluoride, Optical Grade
Description
Barium fluoride is used in ceramic flux, carbon brushes for electrical equipment, glass making, in the manufacture of other fluorides, crystals for spectroscopy, electronics and dry-film lubricants. Used for the antiseptic. For industry application, such as electron, measuring applicance, metallurgy, ceramics. Barium fluoride is often suitable for applications in the passive IR band (8 to 14 ?m) and is often used as a viewport window for thermography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 2260°C |
Melting Point | 1353°C |
Physical Form | Broken Crystals |
Quantity | 25 g |
UN Number | UN1564 |
Merck Index | 14,974 |
Solubility Information | Soluble in methanol,ethanol,water 1.2g/L. |
IUPAC Name | barium(2+) difluoride |
Formula Weight | 175.36 |
Grade | Optical |
Show More |
Safety and Handling
Hazard Category | H301-H332 |
Hazard Statement | GHS H Statement H302-H332 Harmful if swallowed. Harmful if inhaled. |
Precautionary Statement | P261-P264b-P270-P271-P301+P310-P304+P340-P312-P330-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S. |
EINECSNumber | 232-108-0 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.