Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium fluoride, Puratronic™, 99.99% (metals basis)

Catalog No. p-7048473
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 7787-32-8 | BaF2 | 175.32 g/mol

Barium fluoride is used in windows for thermal imaging inspection application in electric power facilities. Barium fluoride can be used to make optical components like lenses, as it is transparent in UV (ultraviolet) and IR (infrared) regions. It is used in windows for IR fuel oil analysis. Barium fluoride is one of the fastest scintillators for the detection of X-rays, and gamma rays. It has been found to be useful for the detection of gamma photons in PET (positron emission tomography), high energy neutrons, alpha and beta particles. Its resistant to high energy radiation is higher than all other optical fluorides. It is employed in metallurgy for refining aluminum, production of welding agents, as a preopacifying agent and in glazing frits production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-32-8
Molecular Formula BaF2
Molecular Weight (g/mol) 175.32
MDL Number MFCD00003450
InChI Key OYLGJCQECKOTOL-UHFFFAOYSA-L
Synonym barium fluoride, baryum fluorure french, baryum fluorure, difluorobarium, barium fluoride, ultra dry, barium ii fluoride, barium fluoride, powder, nickelous sulfate; nickel ii sulfate, barium fluoride trace metals basis
PubChem CID 62670
SMILES [F-].[F-].[Ba++]

Specifications

Boiling Point 2260°C
Melting Point 1353°C
Physical Form Powder
Quantity 25 g
UN Number UN1564
Merck Index 14,974
Solubility Information Slightly soluble in water; soluble in methanol,and ethanol.
IUPAC Name barium(2+);difluoride
Formula Weight 175.36
Percent Purity 99.99%
Grade Puratronic™
Odor Odorless
Refractive Index 1.4741
Density 4.83 g/mL
Assay (metals basis)
Chemical Name or Material Barium fluoride
Show More Show Less
Hazard Category H301-H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P310-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
EINECSNumber 232-108-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.