Barium hydroxide, anhydrous, 94 to 98%, Alfa Aesar™

Manufacturer: Alfa Aesar 12195A1

Quantity 1kg

 View more versions of this product

Catalog No. AA12195A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17194-00-2
    Physical Form Powder
    Assay Percent Range 94 to 98%
    Molecular Formula Ba(OH)2
    Formula Weight 171.36
    MDL Number MFCD00003443
    Chemical Name or Material Barium hydroxide, anhydrous

    Specifications

    Formula Weight 171.36
    Quantity 1kg
    Storage Note 1 Air Sensitive & Hygroscopic
    EINECS Number 241-234-5
    Physical Form Powder
    Assay Percent Range 94 to 98%
    Molecular Formula Ba(OH)2
    CAS 17194-00-2
    MDL Number MFCD00003443
    Chemical Name or Material Barium hydroxide, anhydrous, anhydrous

    Air Sensitive & Hygroscopic

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit