Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Hydroxide, Octahydrate, ACS, 98%, Spectrum™ Chemical
SDP

Catalog No. 18609009 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-609-009 12 kg Poly Bottle
18-609-005 25 g Poly Bottle
18-609-006 125 g Poly Bottle
18-609-007 500 g Poly Bottle
18-609-008 2.5 kg Poly Bottle
18-609-010 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 missing translation for 'options'
Catalog No. 18-609-009 missing translation for 'mfr' Spectrum Chemical Manufacturing Corporation missing translation for 'supplierNo' B104012KG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 12230-71-6
Molecular Formula BaH18O10
Molecular Weight (g/mol) 315.46
MDL Number MFCD00149152
InChI Key ZUDYPQRUOYEARG-UHFFFAOYSA-L
IUPAC Name barium(2+) octahydrate dihydroxide
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]

Specifications

CAS Max % 1
Physical Form Crystals and/or Chunks
Quantity 12 kg
Percent Purity 98%
Grade Reagent
Packaging Poly Bottle
Chemical Name or Material Barium Hydroxide, Octahydrate

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.