Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Hydroxide, Saturated, Fisher Chemical™
SDP

Catalog No. p-212871 Shop All Fisherbrand Products
Click to view available options
Quantity:
250 mL
500 mL
Packaging:
Natural Poly Bottle

Chemical Identifiers

CAS 7732-18-5, 12230-71-6
Molecular Formula BaH18O10
Molecular Weight (g/mol) 315.46
MDL Number MFCD00149152
InChI Key ZUDYPQRUOYEARG-UHFFFAOYSA-L
Synonym barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
PubChem CID 17749109
IUPAC Name barium(2+) octahydrate dihydroxide
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]

Specifications

CAS Min % 91.0, 7.26
CAS Max % 94.0, 7.56
Physical Form Liquid
Quantity 250 mL
Grade Laboratory
Packaging Natural Poly Bottle
Chemical Name or Material Barium Hydroxide, Saturated
DOTInformation DOT Class 8, : Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.