Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium iodide hydrate, 95%

Catalog No. AA1512614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1512614 25 g
AA1512622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1512614 Supplier Thermo Scientific Chemicals Supplier No. 01512614
Add to Cart

Light Sensitive and Hygroscopic | CAS: 13718-50-8 | BaI2 | 391.14 g/mol

Useful in pyrotechnic devices. A reagent for facile synthesis of poly(phenylcarbyne).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13718-50-8
Molecular Formula BaI2
Molecular Weight (g/mol) 391.14
MDL Number MFCD00003452
InChI Key SGUXGJPBTNFBAD-UHFFFAOYSA-L
IUPAC Name barium(2+) diiodide
SMILES [I-].[I-].[Ba++]

Specifications

Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 95%
Solubility Information Freely soluble in water. Soluble in alcohol and acetone
Formula Weight 391.15 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Chemical Name or Material Barium iodide hydrate
Hazard Category H302+H332
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 237-276-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.