Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium metaphosphate, 90+%, Thermo Scientific Chemicals

Catalog No. p-7049777
Click to view available options
Quantity:
500 g

Catalog No. AA2264136

Supplier: Thermo Scientific Chemicals 02264136

Only null left
Add to Cart

Barium metaphosphate, used in glasses, porcelains, and enamels, as a flame retardant and in phosphors applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13762-83-9
Molecular Formula BaO6P2
Molecular Weight (g/mol) 295.27
MDL Number MFCD00061417
InChI Key RYSCSJQMVDRYDV-UHFFFAOYSA-J
Synonym barium metaphosphate, barium 2+ ; dioxido oxo phosphanium, ba.2o3p, bismetaphosphoric acid barium salt, barium 2+ bis trioxidophosphate 1-
PubChem CID 16164194
SMILES [Ba+4].[O-][P]([O-])=O.[O-][P]([O-])=O

Specifications

Melting Point 1560°C
Physical Form Powder
Quantity 500 g
UN Number UN1564
Solubility Information Insoluble in water; Soluble in acids by a slow dissolution
Formula Weight 295.25
Percent Purity ≥90%
Odor Odorless
Chemical Name or Material Barium metaphosphate
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
EINECSNumber 237-362-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.