Barium nitrate, ACS, 99+% Alfa Aesar

Manufacturer: Alfa Aesar 1219436

Quantity 500g

 View more versions of this product

Catalog No. AA1219436

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10022-31-8
    Physical Form Crystalline
    Assay Percent Range >99%
    Molecular Formula Ba(NO3)2
    Formula Weight 261.35
    MDL Number MFCD00003442
    Chemical Name or Material Barium nitrate
    Grade ACS Grade

    Specifications

    Formula Weight 261.35
    Quantity 500g
    Color White
    DOT Information DOT Class: 5.1, Packaging Group: II
    Density 3.24g/cm3
    EINECS Number 233-020-5
    Physical Form Crystalline
    Assay Percent Range >99%
    Appearance White crystalline
    MDL Number MFCD00003442
    CAS 10022-31-8
    Health Hazard 1 H272-H302-H332
    Molecular Formula Ba(NO3)2
    Chemical Name or Material Barium nitrate
    Grade ACS Grade

    H272-H302-H332

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit