Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium oxalate, Puratronic™, 99.999% (metals basis)

Catalog No. p-7049140
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 516-02-9 | C2BaO4 | 225.345 g/mol

Barium oxalate, Puratronic isused as a green pyrotechnic colorant generally in specialized pyrotechnic compositions containing magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 516-02-9
Molecular Formula C2BaO4
Molecular Weight (g/mol) 225.345
MDL Number MFCD00054429
InChI Key GXUARMXARIJAFV-UHFFFAOYSA-L
Synonym barium oxalate, unii-54r8vvf8zk, ethanedioic acid, barium salt 1:1, barium 2+ ; oxalate, barium 2+ ion oxalate, 54r8vvf8zk, oxalic acid barium, barium 2+ oxalate ion, acmc-1b1e7, ethanedioic acid,barium salt 1:1
PubChem CID 68201
IUPAC Name barium(2+);oxalate
SMILES C(=O)(C(=O)[O-])[O-].[Ba+2]

Specifications

Melting Point 400°C (decomposition)
Color White
Physical Form Powder
Quantity 5 g
Linear Formula BaC2O4
UN Number UN1564
Merck Index 14,985
Solubility Information Soluble in water.
Formula Weight 225.36
Percent Purity 99.999%
Grade Puratronic™
Odor Odorless
Density 2.658 g/mL
Assay (metals basis)
Chemical Name or Material Barium oxalate
Show More Show Less
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
EINECSNumber 208-216-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.